Learn More
Medchemexpress LLC Pyrazolo[1,5-a]pyrido[4,3-d]pyrimidin-9(1H)-one derivative, hydrochloride | 1775330-54-5 | 99.9% | 571.78 g·mol⁻¹ | C22H19BrClF3N6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY154912A10MG
Encompass_Preferred
PAT1inh-B01 hydrochloride is a research-grade small-molecule inhibitor selective for SLC26A6 (PAT1), supplied as the hydrochloride salt and intended for in vitro studies of PAT1-mediated anion exchange.
  • Selective inhibitor of SLC26A6 with a reported IC50 of 350 nM.
  • Supplied as a hydrochloride salt suitable for aqueous handling and formulation.
  • High purity (~99.9%) for reliable experimental performance.
  • Well-characterized physicochemical profile (MW 571.78 g·mol⁻¹; formula C22H19BrClF3N6O2).
  • Soluble in DMSO at approximately 10 mg/mL with ultrasonic warming.
  • Available in small research pack sizes for assay development and screening.
  • Store solid at 4°C; follow solvent storage recommendations for long-term stability.
Catalog No. 50-002-86157