Learn More
Medchemexpress LLC Pyrazosulfuron-ethyl | 93697-74-6 | 98.42% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0865
Encompass_Preferred
Pyrazosulfuron-ethyl is an acetolactate synthase inhibiting herbicide belonging to the sulfonylurea family, widely utilized for controlling weed growth in commercial cereal, soybean, and vegetable fields. Research indicates that it inhibits biomass production in *Rhodopseudomonas palustris* PSB-S, alters cell morphology, suppresses flagella formation, and reduces pigment biosynthesis by significantly suppressing carotenoid biosynthesis.
  • Extensively used for controlling weed growth
  • Inhibits acetolactate synthase
  • Belongs to the sulfonylurea family of herbicides
  • Alters cell morphology in certain organisms
  • Suppresses flagella formation and reduces pigment biosynthesis
Catalog No. 50-004-50787