Learn More
Cayman Chemical PyrenophorIn 100ug
SDP
Supplier:  Cayman Chemical 33502100 UG
Encompass_Preferred
A fungal metabolite; inhibits seed germination and reduces root growth of the graminaceous plants A. sterilis, A. fatua, A. sativa, T. aestivum, and H. vulgare at 60 µM; reduces chlorophyll levels in leaf sections from seedlings of the same plants at 320 µM
Catalog No. 50-294-7692