Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR pipettes Class A co1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS7100501EA
Encompass
Tempered top and tip. Red graduation and markings color-coded ASTM E 969.
Catalog No. 50-290-0224