Learn More
Medchemexpress LLC Pyridaben | 96489-71-3 | 98.2% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB081710MM/1ML
Encompass_Preferred
Pyridaben is a mitochondrial electron transport inhibitor (METI) acaricide that promotes the formation of damaging oxygen and nitrogen radicals. It is for research use only.
  • Selectively inhibits complex I (NADH dehydrogenase) with an IC50 value of 2.4 nM.
  • Significantly inhibits rat mitochondrial mtNOS function.
Catalog No. 50-004-08120