Learn More
Medchemexpress LLC Pyridaben | 96489-71-3 | 98.2% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0817100MG
Encompass_Preferred
Pyridaben is a mitochondrial electron transport inhibitor (METI) acaricide that promotes the formation of damaging oxygen and nitrogen radicals. Pyridaben selectively inhibits complex I (NADH dehydrogenase) with an IC50 value of 2.4 nM (assay sites: rat liver and bovine heart mitochondria). Pyridaben also significantly inhibits rat mitochondrial mtNOS function.
  • Mitochondrial electron transport inhibitor (METI) acaricide.
  • Promotes the formation of damaging oxygen and nitrogen radicals.
  • Selectively inhibits complex I (NADH dehydrogenase) with an IC50 value of 2.4 nM (assay sites: rat liver and bovine heart mitochondria).
  • Significantly inhibits rat mitochondrial mtNOS function.
Catalog No. 50-004-08406