Learn More
AdipoGen Pyridafol
SDP
Supplier:  AdipoGen CDXP0133M025
Encompass_Preferred
Chemical. CAS 40020-01-7. Formula C10H7ClN2O. MW 206.63. Synthetic. Pyridazine herbicide. Mode of action is through the inhibition of photosynthetic electron transport at the photosystem II. Used against broad-leaved weeds including nightshade, cleavers, nettles, chickweed and grasses including barnyard grass, crabgrass, foxtail. Compound can be used as analytical reference material.
Catalog No. 50-256-0229