Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridazine-4-carboxylic acid | 50681-25-9 | 124.099 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0020661G

Encompass_Preferred

Pyridazine-4-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Molecular weight of 124.099
  • Purity of 99.64%
  • Off-white to yellow solid appearance
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-47843


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.