Learn More
Matrix Scientific Pyridine-2-boronic acid, 197958-29-5, MFCD00151844, 1g
Supplier: Matrix Scientific 0819981G
Molecular Formula C5H6BNO2, Purity 95%, Molecular Weight 122.92, Melting Point >300°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.