Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridine, 3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-2-nitro- | 877397-70-1 | 99.9% | C13H9Cl2FN2O3 | 1 G
SDP

Supplier:  Medchemexpress LLC HY754681G

Encompass_Preferred

This product is Crizotinib impurity 1, also known as Pyridine, 3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]-2-nitro-. It features a high purity of 99.94% as determined by HPLC.

  • High purity (99.94% HPLC)
  • Powder storage conditions: -20°C for 3 years or 4°C for 2 years
  • In solvent storage conditions: -80°C for 6 months or -20°C for 1 month
  • Appearance: Off-white to light yellow solid

Catalog No. 50-004-34567


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.