Learn More
Medchemexpress LLC Pyridine, 3-methoxy-5-[2-(2-pyridinyl)ethynyl]- | 524924-76-3 | 98.1% | 210.23 g/mol | C13H10N2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY1251010MG
Encompass_Preferred
Methoxy-PEPy is a potent, selective negative allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) supplied for research use. It is a small-molecule solid (C13H10N2O, CAS 524924-76-3) with a molecular weight of approximately 210.23 g/mol and typical purity around 98%. The compound is provided as a dry powder or as a 10 mM solution in DMSO and is intended for receptor pharmacology, in vitro assays, and preclinical studies.
  • Potent mGlu5 antagonist with low-nanomolar activity.
  • High purity suitable for pharmacology studies.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • High solubility in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Intended for research use only.
Catalog No. 50-002-38482