Learn More
AdipoGen Pyridine hydrochloride
SDP
Supplier:  AdipoGen CDXP0710G500
Encompass_Preferred
Chemical. CAS 628-13-7. Formula C5H5N . HCl. MW 115.56. Pyridine hydrochloride can be used as a building block, starting material or reagent in organic synthesis and as a laboratory reagent.
Catalog No. 50-256-0169