Learn More
Medchemexpress LLC Pyrido[3,4-d]pyridazine-8-carbonitrile, 7-[[(1R,2S)-2-aminocyclohexyl]amino]-3,4-dihydro-5-[(3-methylphenyl)amino]-4-o | 2304654-43-9 | 99.8% | 389.45 | 5 MG

Supplier: Medchemexpress LLC 5MGHY128576
This product is a potent and orally active FER tyrosine kinase inhibitor, exhibiting anti-cancer activity with an IC50 of 0.49 nM. It is intended for research use only and not for patient use.
- Potent and orally active FER tyrosine kinase inhibitor
- IC50 of 0.49 nM
- Exhibits anti-cancer activity
- Intended for research use only
- Purity: 99.8%
- Molecular weight: 389.45
- Appearance: White to light yellow solid
- Soluble in DMSO at 200 mg/mL (513.54 mM)
- GI50 of 220 nM for growth inhibition in mouse mock transfected BA/F3 cells
- Long-term storage for powder at -20°C for 3 years, 4°C for 2 years
- Long-term storage in solvent at -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.