Learn More
Medchemexpress LLC Pyrido[4,3-b]indole derivatives and their use as pharmaceuticals | 2765273-11-6 | 99.7% | 341.23 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16054610MM/1ML
Encompass_Preferred
cGAS-IN-2 (compound 109) is a potent inhibitor of Cyclic GMP-AMP Synthase (cGAS), with an IC50 of 0.01512 μM for human cGAS. This product is intended for research use only.
  • Potent inhibitor of cyclic GMP-AMP synthase (cGAS).
  • Available in solid and solution forms.
  • Different unit sizes are available for various research needs.
  • Documentation including data sheet, COA, and SDS are provided.
  • Detailed solubility and storage information for in vitro and in vivo use.
Catalog No. 50-003-91384