Learn More
Medchemexpress LLC Pyridoclax | 1651890-44-6 | 100.0% | 426.51 g/mol | C29H22N4 | 25MG

Supplier: Medchemexpress LLC HY1252725MG
Pyridoclax is a small-molecule inhibitor of the anti-apoptotic protein Mcl-1 intended for research use. It is supplied as a white to off-white solid with high reported purity and is used to study apoptosis and Bcl-2 family signaling. Available as powder and as a DMSO solution; follow recommended storage conditions.
- Mcl-1 inhibitor for apoptosis and Bcl-2 family research.
- High reported purity (99.95%).
- Molecular formula C29H22N4, molecular weight 426.51 g/mol.
- White to off-white solid appearance.
- Available as powder and as a 10 mM solution in DMSO.
- Recommended storage: powder -20°C for long term; in solvent -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.