Learn More
Medchemexpress LLC Pyridostatin hydrochloride | 1781882-65-2 | C31H37Cl5N8O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15176A100MG
Encompass_Preferred
Pyridostatin hydrochloride is a potent G-quadruplex DNA stabilizing agent that targets DNA and RNA G4s within cells. It effectively promotes growth arrest in various human cancer cells by inducing replication- and transcription-dependent DNA damage. This highly selective molecule also specifically targets the proto-oncogene Src, leading to a reduction in SRC protein levels and SRC-dependent cellular motility, particularly in human breast cancer cells.
  • Stabilizes G-quadruplex DNA (Kd=490 nM)
  • Targets DNA and RNA G4s in cellular environments
  • Promotes growth arrest in human cancer cells
  • Induces replication- and transcription-dependent DNA damage
  • Reduces SRC protein levels and cellular motility in breast cancer cells
  • Highly selective G-quadruplex DNA-binding small molecule
  • Induces cell cycle arrest in over 60 cancer cell lines
  • Downregulates the BRCA1 protein transcriptionally
Catalog No. 50-003-74124