Learn More
Cayman Chemical PyrIdostatIntrIfluoROCTAt 10mg
SDP
Supplier:  Cayman Chemical 1801310 MG
Encompass_Preferred
A stabilizer of G-quadruplexes that induces DNA damage and cell cycle arrest (Kd = 490 nM)
Catalog No. 50-296-7452