Learn More
TARGETMOL CHEMICALS INC pyridostigmine 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T86315MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Pyridostigmine(Regonol) is an acetylcholinesterase inhibitor that can treat myasthenia gravis by increasing acetylcholine at the postsynaptic motor endplate. Purity 99.85%
Catalog No. 50-287-7032