Learn More
Apexbio Technology LLC Pyridostigmine Bromide 101-26-8 1g
SDP
Supplier:  Apexbio Technology LLC B16131000
Encompass_Preferred
Pyridostigmine Bromide (CAS 101-26-8) is a synthetic small molecule that functions as a reversible inhibitor of acetylcholinesterase By binding to the active site of the enzyme it impedes the hydrolysis of acetylcholine resulting in enhanced cholinergic transmission at neuromuscular junctions and within the parasympathetic nervous system Pyridostigmine is widely utilized in preclinical and clinical research to investigate cholinergic mechanisms model synaptic transmission and study disorders involving neuromuscular conduction Its application extends to the exploration of therapeutic approaches for diseases such as myasthenia gravis
Catalog No. 50-000-90597