Learn More
Medchemexpress LLC Pyridostigmine bromide | 101-26-8 | 98.86% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0207A5G
Encompass_Preferred
Pyridostigmine bromide is an orally active cholinesterase (ChE) inhibitor used in cardiovascular disease research. It is for research use only.
  • Enhances SH-SY5Y cell activity and mitochondrial activity at low concentrations.
  • Promotes accumulation of SH-SY5Y cells in the S phase at 60-80 ng/mL.
  • Upregulates expression of telomerase gene, p53 gene, and DNMT1 gene in SH-SY5Y cells.
  • Reduces acetylcholinesterase activity in blood in C57BL/6J mice.
  • Improves myocardial changes and reduces inflammatory responses in a chronic Chagas cardiomyopathy mouse model.
Catalog No. 50-004-07035