Learn More
Medchemexpress LLC Pyridostigmine bromide | 101-26-8 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0207A10MM/1ML
Encompass_Preferred
Pyridostigmine bromide is an orally active cholinesterase (ChE) inhibitor supplied as a 10 mM, 1 mL solution in DMSO. It is primarily used in cardiovascular disease research and has demonstrated various biological activities in both in vitro and in vivo studies.
  • Orally active cholinesterase (ChE) inhibitor
  • Enhances SH-SY5Y cell activity and mitochondrial activity at low concentrations
  • Promotes accumulation of SH-SY5Y cells in the S phase
  • Upregulates telomerase, p53, and DNMT1 genes
  • Reduces acetylcholinesterase activity in mice
  • Improves myocardial changes in a Chagas cardiomyopathy mouse model
Catalog No. 50-004-06645