Learn More
Medchemexpress LLC Pyridostigmine bromide | 101-26-8 | MFCD00079283 | 98.86% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0207A500MG
Encompass_Preferred
Pyridostigmine bromide is an orally active cholinesterase (ChE) inhibitor, functioning as a parasympathomimetic and a reversible cholinesterase inhibitor. It is primarily used in cardiovascular disease research and is intended for research purposes only.
  • Orally active cholinesterase inhibitor
  • Functions as a parasympathomimetic
  • Reversible cholinesterase inhibitor
  • Purity of 98.86%
  • Used in cardiovascular disease research
  • Intended for research use only
Catalog No. 50-004-06245