Learn More
Cayman Chemical PyrIdoxal 5-phosphatehydra 1g
SDP
Supplier:  Cayman Chemical 203521 G
Encompass_Preferred
The active form of vitamin B6, which serves as a cofactor for many different enzymes; used to study the pyridoxal 5′-phosphate-dependent active sites of these enzymes
Catalog No. 50-298-4647