Learn More
Medchemexpress LLC Pyridoxal | 66-72-8 | MFCD00464806 | 100.0% | 167.16 | C8H9NO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107469R100MG
Encompass_Preferred
Pyridoxal is the aldehyde form of vitamin B6 supplied as an analytical standard for research and laboratory use. It is intended as a reference material for method development, calibration, and quantitative analysis in biochemical and analytical workflows. The material has a molecular weight of 167.16 and is supplied with supporting documentation for analytical verification.
  • High purity (reported 99.97%).
  • Suitable as a calibration standard for HPLC and spectroscopic assays.
  • Intended for method development and quantitative analysis.
  • Available in small milligram pack sizes including 100 mg.
  • Accompanied by certificate of analysis and safety data sheet.
Catalog No. 50-002-40153