Learn More
Medchemexpress LLC HY-113010 10mg Medchemexpress, 1-Arachidoyl-sn-glycero-3-phosphocholine CAS:108341-80-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11301010MG
Encompass_Preferred
Medchemexpress, HY-114758 50mg Pyridoxal isonicotinoyl hydrazone CAS:737-86-0 Pyridoxal isonicotinoyl hydrazone (PIH) is a lipophilic, tridentate Fe-chelating agent that shows high Fe chelation efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9216