Learn More
Medchemexpress LLC Pyridoxal Standard 10mg | 66-72-8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY107469R10MG
Encompass_Preferred
Pyridoxal (Standard) is the analytical standard of Pyridoxal This product is intended for research and analytical applications Pyridoxal is a neuroprotectant Pyridoxal is one of the main forms of vitamin B6 Pyridoxal is phosphorylated by pyridoxal kinase to pyridoxal phosphate (HY-B1744) Pyridoxal is oxidized by the liver to 4-pyridoxic acid (HY-113493) and excreted in the urine Pyridoxal has shown promise in the study of carpal tunnel syndrome (CTS)[1 [2 [3
Catalog No. 50-002-40340