Learn More
Medchemexpress LLC Pyridoxamine-d3 dihydrochloride | 1173023-45-4 | 99.3% | C8H11D3Cl2N2O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY141923S1MG
Encompass_Preferred
Pyridoxamine-d3 (dihydrochloride) is the deuterium labeled Pyridoxamine dihydrochloride. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-53210