Learn More
Cayman Chemical PyrIdoxamInhydrochlorIde 100g
SDP
Supplier:  Cayman Chemical 33899100 G
Encompass_Preferred
A 4-methylamine form of vitamin B6; scavenges isoketals in cell-free assays and inhibits the formation of lysyl-levuglandin-lactam adducts in dazoxiben-stimulated isolated human platelets at 0.1 and 1 mM; inhibits development of acellular capillaries and accumulation of CML in retina in a rat model of STZ-induced diabetes at 1 g/L in the drinking water; reverses STZ-induced deficits in the novel object recognition task in a rat model of diabetes at 0.4 and 1 g/L in the drinking water
Catalog No. 50-294-8294