Learn More
Abcam Pyrido x atin, MMP-2 inhibitor, 1MG
Supplier:  Abcam AB1443381MG
Encompass_Preferred
MW 263.33 Da, Purity >95%. Free radical scavenger (IC₅₀ = 8 μM). MMP-2 inhibitor. Inhibits lipid peroxidation and DNA synthesis. Shows antifungal effects in vivo (IC₅₀ = 6 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5868