Learn More
Medchemexpress LLC Pyridoxine-d2 hydrochloride | 5027-82-7 | MFCD12022329 | 98.9% | 207.65 g/mol | C8H10D2ClNO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0682S110MG
Encompass_Preferred
Pyridoxine-d2 hydrochloride is the deuterium-labeled form of pyridoxine (vitamin B6) supplied as the hydrochloride salt for research use. It is a pyridine derivative reported to exert antioxidant effects in cell models via the Nrf-2/HO-1 pathway and is used in biochemical and metabolic studies where deuterium labeling aids tracking and quantitation.
  • Deuterium label enables metabolic tracing and mass spectrometry quantitation.
  • High purity (98.9%) suitable for analytical applications.
  • White to off-white solid, stable under recommended storage.
  • Store at 4°C in a sealed container to minimize moisture exposure.
  • Available in milligram-scale quantities for laboratory research.
Catalog No. 50-002-96902