Learn More
U.S. Pharmacopeia Pyridoxine Hydrochloride, 58-56-0, MFCD00012807, 200mg
Supplier:  U.S. Pharmacopeia 1587001
Encompass
Molecular formula C8-H12-Cl-N-O3, Molecular weight 205.64 g/mol, Melting Point 401 - 413.6 °F (205 - 212 °C) (decomposes), Synonyms: Vitamin B6 hydrochloride; Pyridoxol hydrochloride
Catalog No. NC9987932