Learn More
Medchemexpress LLC Pyridoxine (hydrochloride) | 58-56-0 | C8H12ClNO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1505255MG
Encompass_Preferred
Pyridoxine hydrochloride (Pyridoxol; Vitamin B6) is a pyridine derivative. It demonstrates antioxidant effects in a cell model of Alzheimer's disease via the Nrf-2/HO-1 pathway.
  • Exerts antioxidant effects in cell model of Alzheimer's disease via the Nrf-2/HO-1 pathway.
  • Has protective potential against Alzheimer's disease.
  • Attenuates reactive oxygen species levels.
  • Decreases expression of cytoplasmic Nrf2.
  • Upregulates whole-cell HO-1 expression.
Catalog No. 50-003-75632