Learn More
Medchemexpress LLC Pyridoxylamine dihydrochloride | 524-36-7 | MFCD00012808 | 99.1% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1745AR
Encompass_Preferred
Pyridoxylamine dihydrochloride (Standard) is an analytical standard used for research and analytical applications. It functions as an inhibitor of advanced glycation end products (AGEs) and lipoxidation end products (ALEs), providing protection against diabetes-induced retinal vascular lesions.
  • Intended for research and analytical applications
  • Inhibits AGEs and ALEs
  • Protects against diabetes-induced retinal vascular lesions
  • Used as a reference standard in HPLC, GC, and MS experiments
Catalog No. 50-004-14045