Learn More
Medchemexpress LLC Pyrimethanil | 53112-28-0 | 99.85% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB2033500MG
Encompass_Preferred
Pyrimethanil is an anilinopyrimidine and broad-spectrum contact fungicide used for controlling Botrytis spp. on various crops. It inhibits the biosynthesis of methionine and other amino acids in *Botrytis cinerea*. It can be used for research on preventing fungal diseases on fruit, vegetable, and ornamental plants with mold infection.
  • Broad-spectrum contact fungicide
  • Inhibits methionine biosynthesis
  • Suitable for fungal disease prevention research
Catalog No. 50-004-11100