Learn More
Medchemexpress LLC Pyrimethanil | 53112-28-0 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB203310MM/1ML
Encompass_Preferred
Pyrimethanil is an anilinopyrimidine and broad-spectrum contact fungicide used for controlling Botrytis spp. on various crops. It inhibits the biosynthesis of methionine and other amino acids in Botrytis cinerea. It can be used for research on fungal disease prevention in fruit, vegetable, and ornamental plants with mold infection.
  • Broad-spectrum contact fungicide
  • Controls Botrytis spp. on various crops
  • Inhibits biosynthesis of methionine and other amino acids in Botrytis cinerea
  • Can be used for research on fungal disease prevention
  • Decreases polygalacturonase, cellulase, proteinase, and laccase activities
Catalog No. 50-004-11823