Learn More
Medchemexpress LLC Pyrimethanil | 53112-28-0 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB2033100MG
Encompass_Preferred
Pyrimethanil is an anilinopyrimidine and broad-spectrum contact fungicide designed for controlling *Botrytis spp.* across various crops. It functions by inhibiting the biosynthesis of methionine and other amino acids in *Botrytis cinerea*. This product is suitable for research aimed at preventing fungal diseases, such as mold infections, in fruit, vegetable, and ornamental plants.
  • An anilinopyrimidine and broad-spectrum contact fungicide.
  • Effective against *Botrytis spp.* on a wide variety of crops.
  • Inhibits the biosynthesis of methionine and other amino acids in *Botrytis cinerea*.
  • Can be used for research on fungal disease prevention in fruit, vegetable, and ornamental plants with mold infection.
Catalog No. 50-004-11212