Learn More
Cayman Chemical PyrIntegrIn 10mg
SDP
Supplier:  Cayman Chemical 1617910 MG
Encompass_Preferred
A 2,4-disubstituted pyrimidine that, at 2 µM, enhances the survival of hESC more than 30-fold after trypsin-mediated dissociation; increases integrin-dependent attachment of hESC to extracellular matrices
Catalog No. 50-297-2880