Learn More
Medchemexpress LLC HY-13306 5mg Medchemexpress, Pyrintegrin CAS:1228445-38-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133065MG
Encompass_Preferred
Medchemexpress, HY-13306 5mg Pyrintegrin CAS:1228445-38-2 Pyrintegrin is an β1-integrin agonist and a 2,4-disubstituted pyrimidine that promotes embryonic stem cells survival. Pyrintegrin enhances cell-extracellular matrix (ECM) adhesion-mediated integrin signaling. Pyrintegrin can be used as a podocyte-protective agent and has robustly adipogenic[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0277