Learn More
Medchemexpress LLC Pyriproxyfen | 95737-68-1 | 99.4% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB2031R
Encompass_Preferred
Pyriproxyfen is a juvenile hormone analog that prevents larvae from developing into adulthood, rendering them unable to reproduce. It is a pyridine-based pesticide effective against various arthropoda. It is suitable for laboratory centrifugation and storage.
  • Prevents larvae from developing into adulthood.
  • Renders larvae unable to reproduce.
  • Effective against a variety of arthropoda.
  • Solid appearance.
  • White to off-white color.
Catalog No. 50-004-14358