Learn More
Abcam Pyripyropene A, ACAT2 inhibitor, 1MG
Supplier:  Abcam AB1443391MG
Encompass_Preferred
MW 581.6 Da, Purity >95%. Potent, selective ACAT2 inhibitor (IC₅₀ = 70 nM). Shows antihypercholesterolemic and antiatherosclerotic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5866