Learn More
Lonza Walkersville Lonza Monocyte Activation Test System, Human Serum, Rapid, 288 Tests
Catalog No. NC3986805
Encompass
The Monocyte Activation Test – Human Serum Rapid System is a comprehensive kit for pyrogen testing. It includes cryopreserved, pooled Peripheral Blood Mononuclear Cells, Human Serum Supplement, and the PeliKine Human IL-6 ELISA Rapid Kit, enabling 288 tests across three 96-well microtiter plates.
  • Regulatory compliant, detecting full range of pyrogens.
  • Optimized components for robust assay performance.
  • Qualified, cryopreserved cell pools ensure reproducible results.
  • Sustainable system, avoiding animal-based resources.
  • Sensitive detection of non-endotoxin pyrogens (NEP).
  • Recommended for blood-based products.
  • Compatible with appropriate hardware, reagents, and accessories.
  • Test sensitivity for the human serum-based assay is ≤ 0.08.
Catalog No. NC3986805 Supplier Lonza Walkersville Supplier No. 00296408
The Monocyte Activation Test – Human Serum Rapid System is a comprehensive kit for pyrogen testing. It includes cryopreserved, pooled Peripheral Blood Mononuclear Cells, Human Serum Supplement, and the PeliKine Human IL-6 ELISA Rapid Kit, enabling 288 tests across three 96-well microtiter plates.
  • Regulatory compliant, detecting full range of pyrogens.
  • Optimized components for robust assay performance.
  • Qualified, cryopreserved cell pools ensure reproducible results.
  • Sustainable system, avoiding animal-based resources.
  • Sensitive detection of non-endotoxin pyrogens (NEP).
  • Recommended for blood-based products.
  • Compatible with appropriate hardware, reagents, and accessories.
  • Test sensitivity for the human serum-based assay is ≤ 0.08.