Learn More
Lonza Walkersville PYROGENT-5000 Kinetic Turbidimetric LAL Assay, 100 Test Kit
Supplier:  Lonza Walkersville N383
Encompass
Content:2 x 50 tests/vial lysate, 2 vials reconstitution buffer, 1 vial endotoxin
Catalog No. NC9729072