Learn More
Lonza Walkersville PYROGENT-5000 Bulk Kinetic Turbidimetric LAL Assay, 2250 Test Kit
Supplier:  Lonza Walkersville N588
Encompass
The PYROGENT–5000 Kinetic Turbidimetric LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent, placed in the incubating absorbance plate reader, and automatically monitored over time (via WinKQCL Software) for the appearance of turbidity. In the presence of endotoxin, the lysate will begin to gel, causing the solution to become cloudy or turbid. The time required for the change is inversely proportional to the amount of endotoxin present. The concentration in unknown samples can be calculated from a standard curve. The PYROGENT–5000 Kinetic Turbidimetric Assay is perfect for laboratories needing to process large numbers of samples.Benefits:Sensitivity range from 0.01 to 100 EU/mlApplications:Cost-effective method for water and large volume parenterals testingContent:45 x 50 tests/vial lysate45 vials reconstitution buffer10 vials endotoxin
Catalog No. NC0583862
Only null left
Add to Cart
Request a Quote