Learn More
Medchemexpress LLC Pyronaridine tetraphosphate | 76748-86-2 | 99.57% | 910.03 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY14749A500MG
Encompass_Preferred
Pyronaridine tetraphosphate is an orally active Mannich base anti-malarial agent. It is active against P. falciparum and Echinococcus granulosus infection. In vitro, it exhibits anti-P. falciparum activity with IC50 values in the nanomolar range (1.53-3.94 nM). In vivo, it has been shown to reduce parasitic burden in Echinococcus granulosus-infected mice and secondarily infected (cysts) mice, with higher exposure observed in the liver compared to plasma in male ICR mice.
  • Orally active.
  • Mannich base structure.
  • Demonstrates anti-malarial properties.
  • Effective against P. falciparum.
  • Effective against Echinococcus granulosus infection.
  • Reduces parasitic load in animal models.
  • Exhibits preferential distribution to the liver in animal studies.
Catalog No. 50-003-62559