Learn More
Abcam Pyronin B, Cationic dye, 25G
Supplier:  Abcam AB14626125G
Encompass_Preferred
MW 521.11 g/mol. Cationic dye used in the methyl green-pyronin method for differential coloration of nucleic acids.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4787