Learn More
Selleck Chemical LLC Pyrotinib dimaleate S8852-25MG
SDP
Supplier:  Selleck Chemical LLC S885225MG
Encompass_Preferred
Pyrotinib dimaleate is a potent and selective irreversible dual tyrosine kinase inhibitor of EGFR and HER2 with IC50 of 0 013 M and 0 038 M respectively
Catalog No. 50-000-25344