Learn More
Medchemexpress LLC Pyrotinib (racemate) | 1246089-97-3 | >98.0% | 583.08 g/mol | C32H31ClN6O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY104065A25MG
Encompass_Preferred
(Rac)-Pyrotinib is the racemic form of pyrotinib, an irreversible small-molecule inhibitor that targets EGFR and HER2 tyrosine kinases. Supplied as a research-grade solid, it is intended for preclinical studies such as cell-based assays, mechanism-of-action research, and pharmacology investigations.
  • Potent dual EGFR/HER2 inhibitory activity
  • Racemic form useful for stereochemistry and comparative studies
  • High purity for research applications (>98%)
  • Solid formulation for straightforward storage and handling
  • Suitable for cell-based assays and preclinical pharmacology
  • Molecular formula C32H31ClN6O3; molecular weight 583.08 g/mol
Catalog No. 50-002-43914