Learn More
Medchemexpress LLC HY-104065A 5mg Medchemexpress, Pyrotinib Racemate CAS:1246089-97-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104065A5MG
Encompass_Preferred
Medchemexpress, HY-104065A 5mg Pyrotinib Racemate CAS:1246089-97-3 Pyrotinib Racemate is the racemate of Pyrotinib. Pyrotinib is a potent and selective EGFR/HER2 dual inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8200