Learn More
Matrix Scientific Pyrrole-2-carboxylic acid, 634-97-9, MFCD00005219,5g
Supplier: Matrix Scientific 0749855G
Molecular Formula C5H5NO2, Puity 95+%, Molecular Weight 111.10
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.