Learn More
Medchemexpress LLC Pyrrolidin-3-ol-d5 | 1219805-52-3 | 92.15 g/mol | C4H4D5NO | 1 MG

Supplier: Medchemexpress LLC HYW042206S1MG
Pyrrolidin-3-ol-d5 is the deuterium-labeled isotopologue of pyrrolidin-3-ol supplied for research use as a tracer or internal standard in analytical and synthetic chemistry.
- Deuterium-labeled pyrrolidin-3-ol for tracer studies.
- Suitable as an internal standard for mass spectrometry and NMR.
- Available in small research sizes (1 mg, 5 mg, 10 mg), larger sizes by quote.
- Chemical formula C4H4D5NO; molecular weight 92.15 g/mol.
- Storage: pure form -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.